Skip to Content
Merck
All Photos(1)

Key Documents

528781

Sigma-Aldrich

Methyl 3-methoxy-4-methylbenzoate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)CO2CH3
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Assay

96%

mp

50-54 °C (lit.)

SMILES string

COC(=O)c1ccc(C)c(OC)c1

InChI

1S/C10H12O3/c1-7-4-5-8(10(11)13-3)6-9(7)12-2/h4-6H,1-3H3

InChI key

LLEXCSBUSVRBCA-UHFFFAOYSA-N

General description

Methyl 3-methoxy-4-methylbenzoate can be prepared by reacting diazomethane with 3-methoxy-4-methylbenzoic acid in ether.[1]

Packaging

Methyl 3-methoxy-4-methylbenzoate may be used in the synthesis of 3-methoxy-4-methylbenzyl alcohol[1] and zafirlukast.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Zafirlukast.
Wei LI and Qi N.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 8, 001-001 (2004)
Synthesis of (?)-heritol, a sesquiterpene lactone belonging to the aromatic cadinane group.
Irie H, et al.
Chemical & Pharmaceutical Bulletin, 38(7), 1852-1856 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service