Skip to Content
Merck
All Photos(1)

Key Documents

527955

Sigma-Aldrich

Methanesulfonyl chloride

≥99.7%

Synonym(s):

Mesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2Cl
CAS Number:
Molecular Weight:
114.55
Beilstein:
506297
EC Number:
MDL number:
PubChem Substance ID:

Assay

≥99.7%

availability

available only in EU

refractive index

n20/D 1.452 (lit.)

bp

60 °C/21 mmHg (lit.)

density

1.48 g/mL at 25 °C (lit.)

SMILES string

CS(Cl)(=O)=O

InChI

1S/CH3ClO2S/c1-5(2,3)4/h1H3

InChI key

QARBMVPHQWIHKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Torisawa et al.
Bioorganic & medicinal chemistry letters, 17(2), 448-452 (2006-10-28)
Continuing search for beneficial additive toward the Beckmann rearrangement (BR) of indanone oxime has revealed that common Lewis acid catalyst in methanesulfonyl chloride (MsCl) showed increasing efficiency in this ionic rearrangement. The new protocol with MsCl is superior to the
P Sipi et al.
Mutation research, 390(1-2), 105-112 (1997-04-24)
The genotoxicity of effluents collected from a conventional 5-stage softwood kraft pulp bleaching process was studied in Chinese hamster ovary (CHO) cells in vitro. Spent liquor from the first chlorination stage (C/D), where elemental chlorine and chlorine dioxide had been
Gas-liquid chromatographic determination of residues of methiocarb and its toxic metabolites with the flame photometric detector after derivatization with methanesulfonyl chloride.
J C Maitlen
Journal of agricultural and food chemistry, 29(2), 260-264 (1981-03-01)
Maurizio Tamba et al.
Journal of the American Chemical Society, 129(28), 8716-8723 (2007-06-22)
The one-electron reduction of methanesulfonyl chloride (MeSO2Cl) leads, in the first instance, to an electron adduct MeSO2Cl(.)(-) which lives long enough for direct detection and decays into sulfonyl radicals MeSO2(.) and Cl(-), with k = 1.5 x 10(6) s(-1). Both
R Kullberg et al.
The Journal of physiology, 374, 413-427 (1986-05-01)
The development of miniature end-plate currents (m.e.p.c.s) was studied in the superior oblique and interhyoideus muscles of Xenopus laevis. An analysis of m.e.p.c. decays shows that each muscle possesses its own characteristic programme of end-plate current development. In the superior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service