Skip to Content
Merck
All Photos(1)

Key Documents

525898

Sigma-Aldrich

Methyl (1-methylindolyl)-3-glyoxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,526.00

R$1,526.00


Check Cart for Availability


Select a Size

Change View
5 G
R$1,526.00

About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,526.00


Check Cart for Availability

Assay

97%

mp

96-100 °C (lit.)

SMILES string

COC(=O)C(=O)c1cn(C)c2ccccc12

InChI

1S/C12H11NO3/c1-13-7-9(11(14)12(15)16-2)8-5-3-4-6-10(8)13/h3-7H,1-2H3

InChI key

SBHIWUQNUXJUMN-UHFFFAOYSA-N

General description

Methyl (1-methylindolyl)-3-glyoxylate can be prepared from 1-methylindole.[1]

Application

  • Reactant for synthesis of sotrastaurin analogs
  • Reactant for preparation of protein kinase C (PKC) inhibitors
  • Reactant for preparation of indolyl diols
Methyl (1-methylindolyl)-3-glyoxylate may be used to synthesize 3-[(1-methyl)-3-indolyl]-4-(3-indolyl)-1H-pyrrole-2,5-dione and 3,4-[(1-methyl)-3-indolyl]-1H-pyrrole-2,5-dione.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New, Efficient Method for the Synthesis of Bisindolylmaleimides.
Margaret M. Faul et al.
The Journal of organic chemistry, 63(17), 6053-6058 (2001-10-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service