Skip to Content
Merck
All Photos(1)

Key Documents

525022

Sigma-Aldrich

4-Aminoindole

97%

Synonym(s):

(Indol-4-yl)amine, 4-Indolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
Beilstein:
114919
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

106-109 °C (lit.)

SMILES string

Nc1cccc2[nH]ccc12

InChI

1S/C8H8N2/c9-7-2-1-3-8-6(7)4-5-10-8/h1-5,10H,9H2

InChI key

LUNUNJFSHKSXGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminoindole is an indole derivative. Its cytokinin activity has been assessed by tobacco pith callus bioassay.[1] 4-Aminoindole can be prepared by reacting 2,6-dinitrotoluene and N,N-dimethylformamide dimethylacetal in anhydrous DMF.[2]

Application

4-Aminoindole may be used to synthesize:
  • macrolactam tumour promoter indolactam V[3]
  • tricyclic structure of 2-substituted-pyrrolo[2,3-h]quinolin-4-one[2]
  • 4-azidoindole[4]
Reactant for preparation of:
  • Inhibitors of bacterial thymidylate synthase
  • Mimetics of non-alkaloid toxin lignan anticancer and antiviral agent Podophyllotoxin (PPT)
  • Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
  • Protein kinase C θ (PKCθ) inhibitors
  • Indolic non-peptidic HIV protease inhibitors
  • Transient receptor potential cation channel subfamily V member 1 (TRPV1) antagonists
  • Cyclooxygenase-2 (COX-2) and lipoxygenase (LOX) inhibitors
  • 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) inhibitors
  • Short chain 4-substituted indoles as potent αvβ3 antagonist
  • Ligands of serotonin transporter and 5-HT1A receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of azidoindoles and azidotryptophans.
Melhado LL and Leonard NJ.
The Journal of Organic Chemistry, 48(25), 5130-5133 (1983)
Synthestic approaches to the teleocidin-related tumour promoters: a total synthesis of (?)-indolactam V.
de Laszlo SE, et al.
Journal of the Chemical Society. Chemical Communications, 4, 344-346 (1986)
Cytokinin activity of azaindene, azanaphthalene, naphthalene, and indole derivatives.
Torigoe Y, et al.
Phytochemistry, 11(5), 1623-1630 (1972)
Maria Grazia Ferlin et al.
Journal of medicinal chemistry, 48(9), 3417-3427 (2005-04-29)
In our search for potential new anticancer drugs, we designed and synthesized a series of tricyclic compounds containing the antimitotic 2-phenylazaflavone chromophore fused to a pyrrole ring in a pyrroloquinoline structure. Compounds 8, 18, 19, 22, 23, 25 and 26

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service