Skip to Content
Merck
All Photos(1)

Key Documents

524956

Sigma-Aldrich

Estrone-2,4,16,16-d4 3-sulfate sodium salt

≥95 atom % D, ≥99% (CP), contains 35% TRIS-d5 as stabilizer

Synonym(s):

Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$15,558.00

R$15,558.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
R$15,558.00

About This Item

Empirical Formula (Hill Notation):
C18D4H17NaO5S
CAS Number:
Molecular Weight:
376.44
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

R$15,558.00


Check Cart for Availability

Request more information

isotopic purity

≥95 atom % D

Assay

≥99% (CP)

contains

35% TRIS-d5 as stabilizer

technique(s)

mass spectrometry (MS): suitable

mp

300 °C (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

[Na+].[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CCc2c([2H])c1OS([O-])(=O)=O

InChI

1S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,18+;/m1./s1/i3D,7D2,10D;

InChI key

VUCAHVBMSFIGAI-IGBMKMNFSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service