Skip to Content
Merck
All Photos(1)

Documents

521221

Sigma-Aldrich

Z-Leu-OH

97% (HPLC)

Synonym(s):

N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-(Phenylmethoxy)carbonyl-L-leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, N-Cbz-L-leucine, Benzyloxycarbonyl-L-leucine, Benzyloxycarbonylleucine, Carbobenzoxyleucine, L-(Carbobenzyloxy)leucine, L-N-(Benzyloxycarbonyl)leucine, NSC 60039, Z-L-Leucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
265.30
Beilstein:
1253861
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97% (HPLC)

optical activity

[α]20/D −17°, c = 2 in ethanol

contains

~10% solvents of crystalline

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.512 (lit.)

density

1 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)C[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17)/t12-/m0/s1

InChI key

USPFMEKVPDBMCG-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Peptide-based preparation of potential antitumor agents
  • Preparation of cyclopropyl peptidomimetics
  • Peptide synthesis

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service