Skip to Content
Merck
All Photos(2)

Key Documents

521132

Sigma-Aldrich

1-Ethynyl-2,4,5-trimethylbenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$521.00

R$521.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$521.00

About This Item

Linear Formula:
HC≡CC6H2(CH3)3
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$521.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

47-51 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(cc1C)C#C

InChI

1S/C11H12/c1-5-11-7-9(3)8(2)6-10(11)4/h1,6-7H,2-4H3

InChI key

FXHLQHXVJWYAGQ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krishna Bahadur Somai Magar et al.
Molecular diversity, 17(4), 679-691 (2013-08-03)
The rhodium(II)-catalyzed reactions of cyclic diazo compounds derived from barbituric acid or thiobarbituric acid with arylacetylenes and styrenes were examined. These reactions provide a rapid synthetic route to the preparation of a variety of novel and diverse furo [2,3-d pyrimidine-2,4-diones

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service