Skip to Content
Merck
All Photos(2)

Key Documents

520802

Sigma-Aldrich

Platinum black

low surface area, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.9%

form

powder

reaction suitability

core: platinum
reagent type: catalyst

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buchi FN, et al. et al.
Polymer Electrolyte Fuel Cell Durability, By Felix N. Buchi, Minoru Inaba, Thomas J. Schmidt null
Feng Gong et al.
Physical chemistry chemical physics : PCCP, 13(39), 17676-17682 (2011-09-13)
Monolayer of PDDA/graphene/PDDA/H(2)PtCl(6) is fabricated on conductive glass using electrostatic layer-by-layer self-assembly technique, which is then converted to graphene/Pt monolayer for use as counter electrode in dye-sensitized solar cell (DSSC). As compared to the sputtered Pt counter electrode, the self-assembled
S. Zhang, et al.,
Journal of Power Sources, 196, 9955-9960 (2011)
M. C. S. Escano, et al.,
The Journal of Physical Chemistry C, 115, 19883-19889 (2011)
Z. Liu, et al.,
ACS Applied Materials & Interfaces null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service