Skip to Content
Merck
All Photos(1)

Key Documents

519774

Sigma-Aldrich

Cyclohexylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NHNH2 · HCl
CAS Number:
Molecular Weight:
150.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

110-114 °C (lit.)

functional group

hydrazine

SMILES string

Cl[H].NNC1CCCCC1

InChI

1S/C6H14N2.ClH/c7-8-6-4-2-1-3-5-6;/h6,8H,1-5,7H2;1H

InChI key

JZRHODNPRNTXKO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A variation of the Fischer indolization involving condensation of quinone monoketals and aliphatic hydrazines.
Jinzhu Zhang et al.
Angewandte Chemie (International ed. in English), 52(6), 1753-1757 (2013-01-04)
Phosphorescent Iridium (III) Complexes of Cyclometalated 5-Aryl-1H-1, 2, 4-Triazole Ligands: Structural, Computational, Spectroscopic, and Device Studies.
Dobbs KD, et al.
The Journal of Physical Chemistry C, 118(48), 27763-27771 (2014)
The Reaction of Ethyl Azodicarboxylate with Conjugated Dienes1.
Gillis BT and Beck PE.
The Journal of Organic Chemistry, 27(6), 1947-1951 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service