Skip to Content
Merck
All Photos(1)

Key Documents

515523

Sigma-Aldrich

Ethyl 4-hydroxy-2-quinolinecarboxylate

96%

Synonym(s):

Kynurenic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,175.00

R$1,175.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,175.00

About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,175.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

216-218 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc(O)c2ccccc2n1

InChI

1S/C12H11NO3/c1-2-16-12(15)10-7-11(14)8-5-3-4-6-9(8)13-10/h3-7H,2H2,1H3,(H,13,14)

InChI key

UACLREXZGKWWIC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Jenkins et al.
British journal of pharmacology, 162(3), 733-748 (2010-10-21)
GPR35 is a poorly characterized G protein-coupled receptor at which kynurenic acid has been suggested to be the endogenous ligand. We wished to test this and develop assays appropriate for the study of this receptor. Human and rat orthologues of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service