Skip to Content
Merck
All Photos(3)

Key Documents

515493

Sigma-Aldrich

5,5′-Dibromo-2,2′-bithiophene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4Br2S2
CAS Number:
Molecular Weight:
324.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

144-146 °C (lit.)

SMILES string

Brc1ccc(s1)-c2ccc(Br)s2

InChI

1S/C8H4Br2S2/c9-7-3-1-5(11-7)6-2-4-8(10)12-6/h1-4H

InChI key

SXNCMLQAQIGJDO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Capozzi et al.
Journal of the American Chemical Society, 136(29), 10486-10492 (2014-07-09)
We have measured the single-molecule conductance of a family of oligothiophenes comprising 1-6 thiophene moieties terminated with methyl-sulfide linkers using the scanning tunneling microscope-based break-junction technique. We find an anomalous behavior: the peak of the conductance histogram distribution does not

Articles

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service