Skip to Content
Merck
All Photos(1)

Key Documents

513237

Sigma-Aldrich

1,3,-Dichloro-4,6-dinitrobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)2
CAS Number:
Molecular Weight:
237.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

101-104 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(c(Cl)cc1Cl)[N+]([O-])=O

InChI

1S/C6H2Cl2N2O4/c7-3-1-4(8)6(10(13)14)2-5(3)9(11)12/h1-2H

InChI key

ZPXDNSYFDIHPOJ-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingjie Qin et al.
Journal of medicinal chemistry, 53(13), 4906-4916 (2010-06-10)
By using structure-based drug design and isosteric replacement, diarylaniline and 1,5-diarylbenzene-1,2-diamine derivatives were synthesized and evaluated against wild type HIV-1 and drug-resistant viral strains, resulting in the discovery of diarylaniline derivatives as a distinct class of next-generation HIV-1 non-nucleoside reverse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service