Skip to Content
Merck
All Photos(1)

Key Documents

511609

Sigma-Aldrich

Aluminum-tri-sec-butoxide

99.99% trace metals basis

Synonym(s):

Aluminum sec-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al[OCH(CH3)C2H5]3
CAS Number:
Molecular Weight:
246.32
Beilstein:
3910908
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

liquid

autoignition temp.

763 °F

expl. lim.

9.8 %

reaction suitability

core: aluminum

bp

200-206 °C/30 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

CCC(C)O[Al](OC(C)CC)OC(C)CC

InChI

1S/3C4H9O.Al/c3*1-3-4(2)5;/h3*4H,3H2,1-2H3;/q3*-1;+3

InChI key

WOZZOSDBXABUFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Has been utilized in making high-surface-area alumina using 2,4-pentanedione (Aldrich product P775-4) as a chelating agent.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark B Shiflett et al.
The journal of physical chemistry. B, 110(29), 14436-14443 (2006-07-21)
In this article, we investigate vapor-liquid-liquid equilibria (VLLE) of binary systems using a simple volumetric method. Being different from the usual cloud-point method for the determination of liquid-liquid separation boundaries, the present volumetric method is able to determine the direct
Dagmara Wojtków et al.
The journal of physical chemistry. A, 110(36), 10552-10557 (2006-09-08)
The effect of temperature and concentration on the structure of sec-butyl alcohol and isobutyl alcohol/water binary mixtures in the alcohol-rich region (mole fraction of water X(H2O) < 0.3) has been studied using Fourier transform (FT) near-infrared (NIR) spectroscopy. The experimental
Ravi K Pathak et al.
Environmental science & technology, 46(21), 11660-11669 (2012-09-19)
Limonene has a strong tendency to form secondary organic aerosol (SOA) in the atmosphere and in indoor environments. Initial oxidation occurs mainly via ozone or OH radical chemistry. We studied the effect of O(3) concentrations with or without a OH
Fengli Yang et al.
Chemical communications (Cambridge, England), 47(15), 4469-4471 (2011-03-04)
A new solid acid, based on tantalum hydroxide, was used to catalyze saccharide dehydration into 5-hydroxymethylfurfural (HMF) with high catalytic activity and excellent stability in a water-2-butanol biphasic system. Furthermore, good results were also obtained from Jerusalem artichoke juice with
Flaminia Rondino et al.
Physical chemistry chemical physics : PCCP, 13(3), 818-824 (2010-12-07)
Diastereomeric adducts between (S)-1-(4-fluorophenyl)-ethanol and R and S 2-butanol, formed by supersonic expansion, have been investigated by means of a combination of mass selected resonant two-photon ionization-spectroscopy and infrared depletion spectroscopy. Chiral recognition is evidenced by the specific spectroscopic signatures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service