Skip to Content
Merck
All Photos(1)

Key Documents

499846

Sigma-Aldrich

Tributyl(3-methyl-2-butenyl)tin

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnCH2CH=C(CH3)2
CAS Number:
Molecular Weight:
359.18
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

90%

refractive index

n20/D 1.488 (lit.)

bp

283 °C (lit.)

density

1.069 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C\C=C(\C)C

InChI

1S/C5H9.3C4H9.Sn/c1-4-5(2)3;3*1-3-4-2;/h4H,1H2,2-3H3;3*1,3-4H2,2H3;

InChI key

XEFRYQPTIWSVGI-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service