Skip to Content
Merck
All Photos(1)

Documents

499188

Sigma-Aldrich

Merrifield′s peptide resin

70-90 mesh, extent of labeling: 1.5-2.0 mmol/g Cl- loading, 1 % cross-linked

Synonym(s):

Chloromethylated polystyrene, Chloromethylated stryene/divinylbenzene copolymer, Chloromethylated styrene-divinylbenzene resin, Copolymer of styrene and divinylbenzene, chloromethylated, Poly(styrene-co-vinylbenzyl chloride-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene, chloromethylated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

1.5-2.0 mmol/g Cl- loading

particle size

70-90 mesh

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

ClCc1ccccc1.C=Cc2ccccc2.C=Cc3ccc(C=C)cc3

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service