Skip to Content
Merck
All Photos(1)

Key Documents

498211

Sigma-Aldrich

Sulfonyl chloride, polymer-bound

100-200 mesh, extent of labeling: 1.5-2.0 mmol/g loading, 1 % cross-linked

Synonym(s):

(-)-chiro-Inositol, L-chiro-Inositol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$1,002.00

R$1,002.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$1,002.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

R$1,002.00


Check Cart for Availability

Request a Bulk Order

crosslinking

1 % cross-linked

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: amine reactive

extent of labeling

1.5-2.0 mmol/g loading

particle size

100-200 mesh

SMILES string

C=Cc1ccccc1.ClS(=O)(=O)c2ccccc2.C=Cc3ccc(C=C)cc3

InChI

1S/ClHO2S/c1-4(2)3/h4H

InChI key

JRZWKDKJYNXNGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reacts with nucleophiles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service