Skip to Content
Merck
All Photos(1)

Key Documents

497924

Sigma-Aldrich

6-Cyanohexylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NC(CH2)6ZnBr
CAS Number:
Molecular Weight:
255.47
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.944 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]CCCCCCC#N

InChI

1S/C7H12N.BrH.Zn/c1-2-3-4-5-6-7-8;;/h1-6H2;1H;/q;;+1/p-1

InChI key

YDXOVAJDARDUOR-UHFFFAOYSA-M

Application

6-Cyanohexylzinc bromide is an organozinc compound, which can be used as a reactant in palladium-catalyzed Negishi cross-coupling reaction to construct carbon-carbon bonds by coupling with organic halides or triflates.[1][2][3]
It can also be used as a reactant to synthesize:
  • Cyanohexyl substituted aryl derivatives by palladium catalyzed coupling reaction with aryl bromides.[2]
  • Methyl 5-(6-cyanohexyl)-3-methylfuran-2-carboxylate, a key intermediate which is used in the preparation of cyclic furfuryl ether via Wittig rearrangement.[3]

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective construction of a β-isopropenyl alcohol moiety at the C (2) and (3) of kallolide A and pinnatin A using a [2, 3] Wittig rearrangement of cyclic furfuryl ethers
Tsubuki M, et al.
The Journal of Organic Chemistry, 68(26), 10183-10186 (2003)
Room-temperature Negishi cross-coupling of unactivated alkyl bromides with alkyl organozinc reagents utilizing a Pd/N-heterocyclic carbene catalyst
Hadei N, et al.
The Journal of Organic Chemistry, 70(21), 8503-8507 (2005)
Mechanochemical Activation of Zinc and Application to Negishi Cross-Coupling
Cao Q, et al.
Angewandte Chemie (International Edition in English), 130(35), 11509-11513 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service