Skip to Content
Merck
All Photos(1)

Key Documents

497134

Sigma-Aldrich

1,6-Hexanediol ethoxylate diacrylate

average Mn ~314, contains 300 ppm MEHQ as inhibitor

Synonym(s):

1,6-Hexanediol ethoxylate diacrylate, Ethoxylated 1,6-hexanediol diacrylate, Ethoxylated hexanediol diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nCH2CH2CH2-]2
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

specific activity

2.0 ethoxy groups/molecule

mol wt

average Mn ~314

contains

300 ppm MEHQ as inhibitor

refractive index

n20/D 1.461 (lit.)

density

1.01 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC(=O)OCCOCCCCCCOCCOC(=O)C=C

InChI

1S/C16H26O6/c1-3-15(17)21-13-11-19-9-7-5-6-8-10-20-12-14-22-16(18)4-2/h3-4H,1-2,5-14H2

InChI key

GIMNDOUZTMKUHI-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service