Skip to Content
Merck
All Photos(1)

Key Documents

496898

Sigma-Aldrich

1-Benzyl-2-piperidone

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

refractive index

n20/D 1.553 (lit.)

bp

147 °C/0.125 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCN1Cc2ccccc2

InChI

1S/C12H15NO/c14-12-8-4-5-9-13(12)10-11-6-2-1-3-7-11/h1-3,6-7H,4-5,8-10H2

InChI key

MLEGMEBCXGDFQT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A direct synthesis of [(tert-butoxycarbonyl) methylidene] azacycloalkanes.
Yamaguchi M and Hirao I.
The Journal of Organic Chemistry, 50(11), 1975-1977 (1985)
Debenzylation of N-Benzyl-acylamides.
Sugasawa S andFujii T.
Chemical & Pharmaceutical Bulletin, 6(6), 587-590 (1958)
Thermochemistry of some derivatives of 2-and 4-piperidone.
Ribeiro da Silva MAV and Cabral JI.
Journal of Chemical and Engineering Data, 51(5), 1556-1561 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service