Skip to Content
Merck
All Photos(1)

Documents

496294

Sigma-Aldrich

[2,3,3,3-Tetrafluoro-2-(trifluoromethyl)propyl]oxirane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F7O
CAS Number:
Molecular Weight:
226.09
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.32 (lit.)

bp

81 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(CC1CO1)C(F)(F)F

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.1 °F - closed cup

Flash Point(C)

21.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service