Skip to Content
Merck
All Photos(1)

Documents

495522

Sigma-Aldrich

Cesium fluoroaluminate

−200 mesh, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs2AlF5
CAS Number:
Molecular Weight:
387.78
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.9%

particle size

−200 mesh

mp

423-436 °C (lit.)

density

3.7 g/mL at 25 °C (lit.)

SMILES string

[Cs+].F[Al-](F)(F)F

InChI

1S/Al.Cs.4FH/h;;4*1H/q+3;+1;;;;/p-4

InChI key

UYFXWCIZFDKSTJ-UHFFFAOYSA-J

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service