Skip to Content
Merck
All Photos(1)

Key Documents

495131

Sigma-Aldrich

Aluminum phenoxide

99.9% trace metals basis

Synonym(s):

Aluminum phenate, Aluminum phenolate, Aluminum triphenolate, Aluminum triphenoxide, Triphenoxyaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3Al
CAS Number:
Molecular Weight:
306.29
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

reaction suitability

core: aluminum

mp

156 °C (dec.) (lit.)

SMILES string

O([Al](Oc1ccccc1)Oc2ccccc2)c3ccccc3

InChI

1S/3C6H6O.Al/c3*7-6-4-2-1-3-5-6;/h3*1-5,7H;/q;;;+3/p-3

InChI key

OPSWAWSNPREEFQ-UHFFFAOYSA-K

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service