Skip to Content
Merck
All Photos(1)

Key Documents

494224

Sigma-Aldrich

Tetrabromohydroquinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Br4-1,4-(OH)2
CAS Number:
Molecular Weight:
425.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

252-255 °C (lit.)

SMILES string

Oc1c(Br)c(Br)c(O)c(Br)c1Br

InChI

1S/C6H2Br4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11/h11-12H

InChI key

DTFQULSULHRJOA-UHFFFAOYSA-N

General description

Tetrabromohydroquinone (TBHQ), also known as 2,3,5,6-tetrabromohydroquinone, can be synthesized by reacting bromanil with phenyl phosphine. It crystallizes in the monoclinic space group, P21/n. The effect of C-Br group on the crystal structure of TBHQ has been investigated. Its role in causing green luminescence in Ptychodera flava has been investigated.

Application

Tetrabromohydroquinone may be used in the synthesis of thermoresponsive branched oligoethylene glycol dendrimers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study of the crystal structures of tetrahalogenated hydroquinones and ?-hydroquinone.
Thalladi VR, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 55(6), 1005-1013 (1999)
Mona A Abdel-Rahman et al.
European journal of medicinal chemistry, 69, 848-854 (2013-10-15)
Three interesting thermoresponsive branched oligoethylene glycol dendrimers based on tetrabromohydroquinone were efficiently synthesized from tetrabromohydroquinone and three different oligoethylene glycol derivatives. By visual inspection, all these dendrimers are water-soluble at room temperature. The thermoresponsive behaviors were investigated by using UV/vis
Akira Kanakubo et al.
Luminescence : the journal of biological and chemical luminescence, 20(6), 397-400 (2005-06-21)
2,3,5,6-Tetrabromohydroquinone was isolated as a luminous substance from Ptychodera flava. This compound emitted light after addition of hydrogen peroxide under basic conditions. Since hydroquinone had no fluorescence, further investigation by spectral analysis revealed that riboflavin was the only possible light
Reaction of phenylphosphine with p-quinones.
Pudovik AN, et al.
Russian Chemical Bulletin, 27(7), 1450-1452 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service