Skip to Content
Merck
All Photos(1)

Key Documents

493155

Sigma-Aldrich

Stearic acid-2,2-d2

98 atom % D

Synonym(s):

Octadecanoic acid-2,2-d2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$3,515.00

R$3,515.00


Estimated to ship onApril 17, 2025


Request more information

Select a Size

Change View
1 G
R$3,515.00

About This Item

Linear Formula:
CH3(CH2)15CD2CO2H
CAS Number:
Molecular Weight:
286.49
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

R$3,515.00


Estimated to ship onApril 17, 2025


Request more information

isotopic purity

98 atom % D

Quality Level

form

solid

bp

361 °C (lit.)

mp

68-70 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(CCCCCCCCCCCCCCCC)C(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i17D2

InChI key

QIQXTHQIDYTFRH-FBCWWBABSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service