All Photos(1)
About This Item
Linear Formula:
BrC6H4CH(OC2H5)2
CAS Number:
Molecular Weight:
259.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
refractive index
n20/D 1.512 (lit.)
bp
95 °C/0.5 mmHg (lit.)
density
1.271 g/mL at 25 °C (lit.)
SMILES string
CCOC(OCC)c1cccc(Br)c1
InChI
1S/C11H15BrO2/c1-3-13-11(14-4-2)9-6-5-7-10(12)8-9/h5-8,11H,3-4H2,1-2H3
InChI key
KDHRJLQXMOBXRV-UHFFFAOYSA-N
General description
3-Bromobenzaldehyde diethyl acetal is an acetal derivative of 3-bromobenzaldehyde.
Application
3-Bromobenzaldehyde diethyl acetal may be used in the preparation of 3-(tris[2-perfluorohexylethyl]stannyl)benzaldehyde.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
219.2 °F - closed cup
Flash Point(C)
104 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Preparation and evaluation of reagents for tagging amino and thiol groups with fluorous stannanes. A convenient method for producing radioiodinated compounds in high effective specific activity.
Donovan AC and Valliant JF
Journal of Labelled Compounds & Radiopharmaceuticals, 54(2), 65-71 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service