Skip to Content
Merck
All Photos(1)

Key Documents

490725

Sigma-Aldrich

Sodium pyruvate-2-13C

99 atom % 13C

Synonym(s):

Pyruvic acid-2-13C sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$4,281.00
1 G
R$5,919.00

R$4,281.00


Estimated to ship onApril 15, 2025


Request more information

Select a Size

Change View
500 MG
R$4,281.00
1 G
R$5,919.00

About This Item

Linear Formula:
CH313COCO2Na
CAS Number:
Molecular Weight:
111.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

R$4,281.00


Estimated to ship onApril 15, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

>300 °C (lit.)

mass shift

M+1

storage temp.

2-8°C

SMILES string

[Na+].C[13C](=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i2+1;

InChI key

DAEPDZWVDSPTHF-CGOMOMTCSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service