Skip to Content
Merck
All Photos(1)

Key Documents

487570

Sigma-Aldrich

Cyanuric chloride-13C3

99 atom % 13C

Synonym(s):

2,4,6-Trichloro-1,3,5-triazine-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$8,375.00

R$8,375.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
R$8,375.00

About This Item

Empirical Formula (Hill Notation):
13C3Cl3N3
CAS Number:
Molecular Weight:
187.39
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

R$8,375.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

bp

190 °C (lit.)

mp

145-147 °C (lit.)

mass shift

M+3

SMILES string

Cl[13c]1n[13c](Cl)n[13c](Cl)n1

InChI

1S/C3Cl3N3/c4-1-7-2(5)9-3(6)8-1/i1+1,2+1,3+1

InChI key

MGNCLNQXLYJVJD-VMIGTVKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service