Skip to Content
Merck
All Photos(2)

Key Documents

484334

Sigma-Aldrich

(1R,2R)-(−)-N-p-Tosyl-1,2-diphenylethylenediamine

greener alternative

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,096.00

R$1,096.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,096.00

About This Item

Linear Formula:
CH3C6H4SO2NHCH(C6H5)CH(C6H5)NH2
CAS Number:
Molecular Weight:
366.48
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

R$1,096.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

optical activity

[α]20/D −35°, c = 1 in chloroform

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

128-131 °C (lit.)

functional group

amine
phenyl

greener alternative category

SMILES string

Cc1ccc(cc1)S(=O)(=O)N[C@@H]([C@H](N)c2ccccc2)c3ccccc3

InChI

1S/C21H22N2O2S/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17/h2-15,20-21,23H,22H2,1H3/t20-,21-/m1/s1

InChI key

UOPFIWYXBIHPIP-NHCUHLMSSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.s

Application

Chiral diamine ligand for cooperative metal-Bronsted acid catalyzed greener reductive amination using hydrogen gas.

Metal-Brønsted Acid Cooperative Catalysis for Asymmetric Reductive Amination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service