Skip to Content
Merck
All Photos(1)

Key Documents

481823

Sigma-Aldrich

Mercury(II) thiocyanate

99.99% trace metals basis

Synonym(s):

Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
Beilstein:
3561344
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

technique(s)

NMR: suitable

impurities

≤110 ppm Trace Metal Analysis

mp

165 °C (dec.) (lit.)

density

3.71 g/mL at 25 °C (lit.)

SMILES string

N#CS[Hg]SC#N

InChI

1S/2CHNS.Hg/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

GBZANUMDJPCQHY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Increasing interest in solid state NMR of heavy-metal nuclei results in the preparation of several Hg-thiocyanate and selenocyanate compounds as potential models.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Koupparis et al.
Journal of pharmaceutical sciences, 75(8), 800-804 (1986-08-01)
An automated flow-injection determination of chloride salts of drugs, based on the colorimetric mercurothiocyanate determination of chloride counterion is described. The majority of these drugs can be determined, using chloride ion standards, with a coefficient of variation less than 1%
Bowmaker, G.A. et al.
Inorganic Chemistry, 37, 1734-1734 (1998)
Ajit Kumar et al.
World journal of microbiology & biotechnology, 31(8), 1211-1216 (2015-05-11)
Xanthobacter autotrophicus GJ10 has been widely studied because of its ability to degrade halogenated compounds, especially 1,2-dichloroethane (1,2-DCA), which is achieved through chromosomal as well as plasmid pAUX1 encoded 1,2-DCA degrading genes. This work described the gene expression and enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service