Skip to Content
Merck
All Photos(1)

Documents

475947

Sigma-Aldrich

4-Iodophenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NCS
CAS Number:
Molecular Weight:
261.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

70-76 °C (lit.)

SMILES string

Ic1ccc(cc1)N=C=S

InChI

1S/C7H4INS/c8-6-1-3-7(4-2-6)9-5-10/h1-4H

InChI key

SNLGBRZZFRSXHA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of some 1, 2, 4-triazole-3 (4h)-thiones obtained from intramolecular cyclization of new 1-(4-(4-x-phenylsulfonyl) benzoyl)-4-(4-iodophenyl)-thiosemicarbazides.
Cretu OD, et al.
J. Serb. Chem. Soc., 75(11), 1463-1471 (2010)
Yu Shi et al.
Analytical chemistry, 81(15), 6438-6448 (2009-07-04)
The exact mass of a peptide differs characteristically from its nominal mass by a value called the mass defect. Limited by possible elemental compositions, the mass defect of peptides has a restricted range, resulting in an unoccupied mass spectral space
Anja Wellejus et al.
Restorative neurology and neuroscience, 30(1), 21-38 (2011-11-16)
Naturally occurring isothiocyanates (ITCs) are known to possess chemopreventive and neuroprotective properties. Our objective was to study the synthetic ITC 4-iodophenyl isothiocyanate (4-IPITC) in different models of neurodegeneration. In vitro, we exposed primary cortical neurons to various insults such as
Xudong Yao et al.
Analytical chemistry, 80(19), 7383-7391 (2008-09-10)
A method termed as the averagine-scaling analysis (ASA) is proposed for predictive design and selection of chemical reagents for modifying peptides, as well as for facile mass spectral analysis of peptide fragment ions with increased mass defects. The ASA method
Alexandre A Oliveira et al.
Dalton transactions (Cambridge, England : 2003), 46(3), 918-932 (2016-12-24)
Complexes [In(2Ac4oClPh)Cl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service