Skip to Content
Merck
All Photos(1)

Key Documents

475416

Sigma-Aldrich

Glycerol, polymer-bound

extent of labeling: 1-2 mmol/g glycerol loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

description

Polymer support is polystyrene

Quality Level

form

solid

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

1-2 mmol/g glycerol loading

storage temp.

−20°C

SMILES string

OCC(O)CO[*]

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service