Skip to Content
Merck
All Photos(1)

Key Documents

474835

Sigma-Aldrich

2,4-Difluorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NCS
CAS Number:
Molecular Weight:
171.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.598 (lit.)

bp

46-48 °C/0.5 mmHg (lit.)

density

1.349 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(N=C=S)c(F)c1

InChI

1S/C7H3F2NS/c8-5-1-2-7(10-4-11)6(9)3-5/h1-3H

InChI key

ABGGPKIFVAIRGU-UHFFFAOYSA-N

General description

2,4-Difluorophenyl isothiocyanate is a fluorinated building block. Its enthalpy of vaporization at boiling point (490.15K) has been reported to be 42.939kjoule/mol.[1]

Application

2,4-Difluorophenyl isothiocyanate may be used in the synthesis of the following 2-(4-(4-X-phenylsulfonyl)benzoyl)-N-(2,4-difluorophenyl)hydrazinecarbothioamides:[2]
  • N-(2,4-difluorophenyl)-2-(4-(phenylsulfonyl)benzoyl)hydrazinecarbothioamide
  • 2-(4-(4-chlorophenylsulfonyl)benzoyl)-N-(2,4-difluorophenyl)hydrazinecarbothioamide
  • 2-(4-(4-bromophenylsulfonyl)benzoyl)-N-(2,4-difluorophenyl)hydrazinecarbothioamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefania-Felicia Barbuceanu et al.
International journal of molecular sciences, 15(6), 10908-10925 (2014-06-19)
In the present investigation, new hydrazinecarbothioamides 4-6 were synthesized by reaction of 4-(4-X-phenylsulfonyl)benzoic acids hydrazides (X=H, Cl, Br) 1-3 with 2,4-difluorophenyl isothiocyanate and further these were treated with sodium hydroxide to obtain 1,2,4-triazole-3-thione derivatives 7-9. The reaction of 7-9 with
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 535-535 (2014)
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service