Skip to Content
Merck
All Photos(1)

Key Documents

473537

Sigma-Aldrich

Tetrakis(ethylmethylamido)titanium(IV)

electronic grade, ≥99.99%

Synonym(s):

Tetrakis(ethylmethylamino)titanium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3C2H5)N]4Ti
CAS Number:
Molecular Weight:
280.28
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Assay

≥99.99%

form

liquid

reaction suitability

core: titanium

bp

80 °C/0.1 mmHg (lit.)

density

0.923 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(C)[Ti](N(C)CC)(N(C)CC)N(C)CC

InChI

1S/4C3H8N.Ti/c4*1-3-4-2;/h4*3H2,1-2H3;/q4*-1;+4

InChI key

LNKYFCABELSPAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 22 Titanium

Application

Precursor to titanium nitride (TiN) thin films by organometallic chemical vapor deposition (OMCVD).[1]

Other Notes

Please inquire for bulk quantity, pricing, and packaging options.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of Materials, 9, 76-76 (1997)

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service