Skip to Content
Merck
All Photos(1)

Key Documents

471615

Sigma-Aldrich

4-Chlorobenzenesulfonyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2NCO
CAS Number:
Molecular Weight:
217.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.558 (lit.)

bp

121 °C/0.5 mmHg (lit.)

density

1.457 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(cc1)S(=O)(=O)N=C=O

InChI

1S/C7H4ClNO3S/c8-6-1-3-7(4-2-6)13(11,12)9-5-10/h1-4H

InChI key

JGHDVROWMPBQSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorobenzenesulfonyl isocyanate is an isocyanate derivative.

Application

4-Chlorobenzenesulfonyl isocyanate may be used as starting reagent in the synthesis of 2-chloroethyl 4-chlorobenzenesulfonyl carbamate and 1-chloro-2-propyl 4-chlorobenzenesulfonyl carbamate.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of sulfonyl isocyanates and sulfonyl isothiocyanates. IX. Routes to substituted oxazolidin-2-ones and oxazolidine-2-thiones.
McFarland JW, et al.
Journal of Heterocyclic Chemistry, 17(2), 271-272 (1980)
Acetylative cleavage of (arylsulfonyl) ureas to N-acetylarenesulfonamides and isocyanates.
Nagasawa HT, et al.
The Journal of Organic Chemistry, 50(24), 4993-4996 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service