Skip to Content
Merck
All Photos(1)

Key Documents

470023

Sigma-Aldrich

D-Threoninol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
105.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D 4.5±0.5°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

57.5-61.5 °C (lit.)

application(s)

peptide synthesis

SMILES string

C[C@H](O)[C@@H](N)CO

InChI

1S/C4H11NO2/c1-3(7)4(5)2-6/h3-4,6-7H,2,5H2,1H3/t3-,4-/m0/s1

InChI key

MUVQIIBPDFTEKM-IMJSIDKUSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Kamiya et al.
International journal of molecular sciences, 21(15) (2020-07-29)
Small interfering RNA (siRNA) has been recognized as a powerful gene-silencing tool. For therapeutic application, chemical modification is often required to improve the properties of siRNA, including its nuclease resistance, activity, off-target effects, and tissue distribution. Careful siRNA guide strand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service