Skip to Content
Merck
All Photos(1)

Key Documents

469505

Sigma-Aldrich

(R)-2-(Boc-amino)-1-propanol

98%, optical purity ee: 98% (GLC)

Synonym(s):

(R)-(+)-2-(tert-Butoxycarbonylamino)-1-propanol, N-Boc-D-alaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

form

solid

optical activity

[α]20/D +11°, c = 1 in chloroform

optical purity

ee: 98% (GLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

58-61 °C (lit.)

application(s)

peptide synthesis

SMILES string

C[C@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C8H17NO3/c1-6(5-10)9-7(11)12-8(2,3)4/h6,10H,5H2,1-4H3,(H,9,11)/t6-/m1/s1

InChI key

PDAFIZPRSXHMCO-ZCFIWIBFSA-N

Application

Used in the synthesis of antithrombotic nipecotamides.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Said Stålsmeden et al.
Organic & biomolecular chemistry, 18(10), 1957-1967 (2020-02-27)
1,4- and 1,5-Disubstituted triazole amino acid monomers have gained increasing interest among peptidic foldamers, as they are easily prepared via Cu- and Ru-catalyzed click reactions, with the potential for side chain variation. While the latter is key to their applicability
X Zheng et al.
Chirality, 7(2), 90-95 (1995-01-01)
The stereoisomers of alpha,alpha'-bis[3-(N,N-diethylcarbamoyl)-piperidino]-p-xylene (1) were synthesized. Rac ethyl nipecotate was resolved by diastereomeric (-)-D- and (+)-L-tartrate salt formation. The enantiomeric esters were hydrolyzed to the corresponding nipecotic acids, which were then converted into t-BOC derivatives. Treatment of the latter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service