Skip to Content
Merck
All Photos(1)

Key Documents

467138

Sigma-Aldrich

Chlorodimethylphenethylsilane

≥98%

Synonym(s):

Chlorodimethyl(2-phenylethyl)silane, Dimethyl(2-phenylethyl)chlorosilane, Dimethyl(phenethyl)chlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
R$995.00

R$995.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
R$995.00

About This Item

Linear Formula:
C6H5CH2CH2Si(CH3)2Cl
CAS Number:
Molecular Weight:
198.76
Beilstein:
2935510
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

R$995.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

bp

57 °C/0.03 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

C[Si](C)(Cl)CCc1ccccc1

InChI

1S/C10H15ClSi/c1-12(2,11)9-8-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3

InChI key

SBBQHOJYUBTWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service