Skip to Content
Merck
All Photos(2)

Key Documents

465798

Sigma-Aldrich

2-tert-Butyl-4-ethylphenol

99%

Synonym(s):

2-tert -Butyl-4-ethylphenol, 4-Ethyl-2-tert -butylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(C2H5)OH
CAS Number:
Molecular Weight:
178.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

bp

250 °C (lit.)

mp

28-32 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C12H18O/c1-5-9-6-7-11(13)10(8-9)12(2,3)4/h6-8,13H,5H2,1-4H3

InChI key

LZHCVNIARUXHAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Mizutani et al.
Toxicology and applied pharmacology, 69(2), 283-290 (1983-06-30)
A comparative test in mice for pulmonary toxicity between butylated hydroxytoluene (2,6-di-tert.-butyl-4-methylphenol, BHT) and 2,6-di-tert.-butyl-4-[alpha, alpha, alpha-2H3]methylphenol (BHT-d3) showed a significantly lower toxic potency of the latter. The rate of in vitro BHT metabolism to 2,6-di-tert.-butyl-4-methylene-2,5-cyclohexadienone (BHT-QM) was slowed by
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service