Skip to Content
Merck
All Photos(1)

Documents

463418

Sigma-Aldrich

Trimethyl(2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl)silane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22Si
CAS Number:
Molecular Weight:
194.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.486 (lit.)

bp

45 °C/0.03 mmHg (lit.)

density

0.852 g/mL at 25 °C (lit.)

SMILES string

CC1=C(C)C(C)=C(C)C1[Si](C)(C)C

InChI

1S/C12H22Si/c1-8-9(2)11(4)12(10(8)3)13(5,6)7/h12H,1-7H3

InChI key

VEUUNIUMKJTBJB-UHFFFAOYSA-N

Application

Trimethyl(2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl)silane may be used for the synthesis of aryl/heteroarylpiperazine derivatives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and preliminary studies of biological activity of amino derivatives of 4-azatricyclo-[5.2. 1.02, 6] dec-8-ene-3, 5-dione with silicon in the structure.
Napiorkowska M, et al.
Heterocyclic Communications, 21(1), 19024-19024 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service