Skip to Content
Merck
All Photos(1)

Key Documents

462128

Sigma-Aldrich

6-Methylpyridine-2-carboxylic acid

Synonym(s):

2-Carboxy-6-methylpyridine, 6-Methylpicolinic acid, NSC 109143, NSC 26023

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

bp

100 °C/4.5 mmHg (lit.)

mp

120-127 °C (lit.)

SMILES string

Cc1cccc(n1)C(O)=O

InChI

1S/C7H7NO2/c1-5-3-2-4-6(8-5)7(9)10/h2-4H,1H3,(H,9,10)

InChI key

LTUUGSGSUZRPRV-UHFFFAOYSA-N

General description

6-Methylpyridine-2-carboxylic acid (2-carboxy-6-methylpyridine, 6-methylpicolinic acid) is a heterocyclic building block.

Application

6-Methylpyridine-2-carboxylic acid may be used in the preparation of 2-(2-benzoxazolyl)-6-methylpyridine and 1,5-cyclooctadiene iridium(I) complexes bearing chelating ligands.
Pharmacological activity studied in regard to:
  • Interactions of carrier ligands of antidiabetic metal complexes with human serum albumin
  • Ability to chelate metalloenzyme inhibitors
  • Interactions of insulin-mimetic zinc(II) complexes with cell constituents

Reactant for:
  • Synthesis of inhibitors of human 11β -hydroxysteroid dehydrogenase type 1
  • Lewis base organocatalysts for stereoselective hydrosilylation reactions

Biological studies of insulin-enhancing complexes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic dehydrogenation of cyclooctane with neutral iridium (I) complexes.
Taubmann S and Alt HG.
Journal of Organometallic Chemistry, 693(10), 1808-1814 (2008)
Rong Gao et al.
Dalton transactions (Cambridge, England : 2003), (41)(41), 5645-5651 (2008-10-16)
A series of new nickel complexes bearing benzoxazolylpyridines was synthesized and characterized by FT-IR spectroscopic and elemental analysis. The molecular structures of two representative complexes were determined by single-crystal X-ray diffraction. The complex [NiCl2[2-(2-benzoxazolyl)-6-methylpyridine]] (5) is a centrosymmetric dinuclear compound
Guo Hai Xu et al.
Acta crystallographica. Section C, Crystal structure communications, 62(Pt 12), m581-m583 (2006-12-07)
The title compound, [CrSn(C6H5)3(C7H6NO2)3Cl][Sn(C6H5)3Cl(CH4O)], was obtained from the reaction of Ph3SnCl with the complex [Cr(C7H6NO2)3] in methanol. The structure contains [Ph3SnCl(MeOH)] (A) and [Ph3SnClCr(C7H6NO2)3] (B) molecules. In molecule A, the Sn atom of Ph3SnCl is coordinated by one methanol molecule.
Ken-Ichi Ohno et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(9), 1017-1021 (2018-08-07)
In this study, proton affinitive derivatization using picolinic acid and its analogs (3- and 6-methylpicolinic acid and 5-butylpicolinic acid) with proton affinitive moieties was performed for the highly sensitive determination of testosterone (T) and 5α-dihydrotestosterone (DHT) in saliva by LC-ESI-MS/MS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service