Skip to Content
Merck
All Photos(2)

Key Documents

461083

Sigma-Aldrich

Di-p-tolylamine

97%

Synonym(s):

4,4′-Dimethyldiphenylamine, 4-Methyl-N -(4-methylphenyl)benzenamine, p ,p ′-Ditolylamine, Bis(4-methylphenyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4)2NH
CAS Number:
Molecular Weight:
197.28
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

78-82 °C (lit.)

SMILES string

Cc1ccc(Nc2ccc(C)cc2)cc1

InChI

1S/C14H15N/c1-11-3-7-13(8-4-11)15-14-9-5-12(2)6-10-14/h3-10,15H,1-2H3

InChI key

RHPVVNRNAHRJOQ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service