Skip to Content
Merck
All Photos(1)

Key Documents

460729

Sigma-Aldrich

DAB-AM-8, Polypropylenimine octaamine Dendrimer, Generation 2.0

volume 922 Å3

Synonym(s):

4,17-Bis(3-aminopropyl)-8,13-bis[3-[bis(3-aminopropyl)amino]propyl]-4,8,13,17-tetraazaeicosane-1,20-diamine, 4,17-Bis-(3-aminopropyl)-8,13-bis-[3-[bis-(3-aminopropyl)-amino]-propyl]-4,8,13,17-tetraazaeicosan-1,20-diamin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2N[(CH2)3N[(CH2)3NH2]2]2]2
CAS Number:
Molecular Weight:
773.28
UNSPSC Code:
12162002

volume

922 Å3

refractive index

n20/D 1.512 (lit.)

pH

12 (25 wt. % in H2O)

viscosity

0.28 poise(50 °C)(lit.)

transition temp

Tg −97 °C

solubility

methanol and water: soluble

density

0.99 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Contains 8 surface primary amino groups.

Application

Coatings, adhesives, plastic additives, cosmetics, catalysts, conductive films, surfactants, controlled release and medical diagnostics.

Legal Information

DSM product

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service