Skip to Content
Merck
All Photos(1)

Key Documents

459542

Sigma-Aldrich

(R)-(−)-4-Benzyl-3-propionyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −102°, c = 1 in ethanol

optical purity

ee: 99% (HPLC)

mp

44-46 °C (lit.)

functional group

phenyl

SMILES string

CCC(=O)N1[C@@H](COC1=O)Cc2ccccc2

InChI

1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m1/s1

InChI key

WHOBYFHKONUTMW-LLVKDONJSA-N

General description

(R)-(−)-4-Benzyl-3-propionyl-2-oxazolidinone is used as a building block in organic synthesis for the preparation of oxazolidinone derivatives.[1][2]

Application

(R)-(−)-4-Benzyl-3-propionyl-2-oxazolidinone can be used as a building block for the preparation of methyl 3-[(S)-3-((R)-4-benzyl-2-oxooxazolidin-3-yl)-2-methyl-3-oxopropyl]benzoate by treating with strong base followed by the addition of methyl 3-bromomethyl benzoate.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxazolidinone cross-alkylation during Evans? asymmetric alkylation reaction
Fresno N, et al.
Tetrahedron, 67(47), 9104-9111 (2011)
A Synthetic Route to β-Hydroxytyrosine-Derived Tetramic Acids: Total Synthesis of the Fungal Metabolite F-14329
Bruckner, S, et al.
Chemistry?A European Journal , 23(24), 5692-5695 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service