Skip to Content
Merck
All Photos(1)

Documents

459305

Sigma-Aldrich

Fmoc-Arg-OH

97%

Synonym(s):

Nα-Fmoc-L-arginine, Nα-(9-Fluorenylmethoxycarbonyl)-L-arginine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N4O4
CAS Number:
Molecular Weight:
396.44
Beilstein:
4828015
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +8°, c = 1 in DMF

impurities

<3% water

mp

145-150 °C (lit.)

SMILES string

NC(=N)NCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H24N4O4/c22-20(23)24-11-5-10-18(19(26)27)25-21(28)29-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2,(H,25,28)(H,26,27)(H4,22,23,24)/t18-/m0/s1

InChI key

DVBUCBXGDWWXNY-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service