Skip to Content
Merck
All Photos(1)

Documents

45885

Sigma-Aldrich

(+)-Cedryl acetate

technical, ≥90% (sum of enantiomers, GC)

Synonym(s):

(3R,3aS,6R,7R,8aS)-Octahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-6-ol-6-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28O2
CAS Number:
Molecular Weight:
264.40
Beilstein:
2052432
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

technical

Assay

≥90% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +26±1°, c = 1% in ethanol

refractive index

n20/D 1.499 (lit.)

bp

>200 °C (lit.)

mp

44-46 °C

density

0.999 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)OC(C)=O

InChI

1S/C17H28O2/c1-11-6-7-13-15(3,4)14-10-17(11,13)9-8-16(14,5)19-12(2)18/h11,13-14H,6-10H2,1-5H3/t11-,13+,14-,16-,17+/m1/s1

InChI key

HQKQRXZEXPXXIG-DTWJZALFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Cedryl acetate is a sesquiterpene compound, which is derived from natural oils found to be volatile in nature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sesquiterpene components of volatile oils as skin penetration enhancers for the hydrophilic permeant 5-fluorouracil.
Cornwell PA and Barry BW
The Journal of Pharmacy and Pharmacology, 46(4), 261-269 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service