Skip to Content
Merck
All Photos(2)

Key Documents

458333

Sigma-Aldrich

Sucrose benzoate

Synonym(s):

Benzoylsaccharose

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$739.00

R$739.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

Select a Size

Change View
500 G
R$739.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

R$739.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

form

solid

Quality Level

optical activity

[α]23/D +35°, c = 1 in chloroform

mp

93-100 °C (lit.)

transition temp

softening point 98 °C (ring and ball)

solubility

aromatics, ketones and esters: soluble
water, alcohols and aliphatics: insoluble

SMILES string

O=C(OC[C@H]1O[C@H](O[C@]3(COC(=O)c2ccccc2)O[C@H](COC(=O)c4ccccc4)[C@@H](OC(=O)c5ccccc5)[C@@H]3OC(=O)c6ccccc6)[C@H](OC(=O)c7ccccc7)[C@@H](OC(=O)c8ccccc8)[C@@H]1OC(=O)c9ccccc9)c%10ccccc%10

InChI

1S/C68H54O19/c69-59(44-25-9-1-10-26-44)77-41-52-54(81-62(72)47-31-15-4-16-32-47)56(83-64(74)49-35-19-6-20-36-49)57(84-65(75)50-37-21-7-22-38-50)67(80-52)87-68(43-79-61(71)46-29-13-3-14-30-46)58(85-66(76)51-39-23-8-24-40-51)55(82-63(73)48-33-17-5-18-34-48)53(86-68)42-78-60(70)45-27-11-2-12-28-45/h1-40,52-58,67H,41-43H2/t52-,53-,54-,55-,56+,57-,58+,67-,68+/m1/s1

InChI key

AKIVKIDZMLQJCH-KWOGCLBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer
Hardening agent for coatings.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service