Skip to Content
Merck
All Photos(1)

Key Documents

457485

Sigma-Aldrich

2-Chloro-5-ethylpyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7ClN2
CAS Number:
Molecular Weight:
142.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.521 (lit.)

bp

223 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

CCc1cnc(Cl)nc1

InChI

1S/C6H7ClN2/c1-2-5-3-8-6(7)9-4-5/h3-4H,2H2,1H3

InChI key

BGLLZQRUXJGTAD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bobby N Glover et al.
The Journal of organic chemistry, 75(11), 3904-3907 (2010-04-30)
A unique buffering effect of various bases, i-Pr(2)NEt and CaCO(3) in particular, was observed for the acid-catalyzed chloro displacement of 2-chloro-5-ethylpyrimidine with a 2-methyl-2-phenylpropanamine. The use of the carefully chosen bases was essential for the progression of the chloro displacement
239. The Dimroth rearrangement. Part I. Some alkylated 2-iminopyrimidines.
Brown DJ and Harper JS.
Journal of the Chemical Society, 1276-1284 (1963)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service