Skip to Content
Merck
All Photos(1)

Key Documents

45720

Sigma-Aldrich

Eserine salicylate salt

≥97.0% (N)

Synonym(s):

Physostigmine salicylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O2 · C7H6O3
CAS Number:
Molecular Weight:
413.47
Beilstein:
3900576
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

biological source

synthetic

Assay

≥97.0% (N)

form

powder

optical activity

[α]20/D −77±2°, c = 1% in ethanol

mp

181-183 °C

SMILES string

OC(=O)c1ccccc1O.CNC(=O)Oc2ccc3N(C)[C@H]4N(C)CC[C@@]4(C)c3c2

InChI

1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13-,15+;/m1./s1

InChI key

HZOTZTANVBDFOF-PBCQUBLHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acetylcholinesterase inhibitor that crosses the blood-brain barrier and forms a carbamylated enzyme complex with acetyl cholinesterase that degrades slowly.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Matschiner et al.
HNO, 49(12), 1026-1028 (2002-01-17)
Appearance of an anticholinergic syndrome after treatment with drugs in therapeutic dosages is seldom reported in the literature. Based on a case report, the development and course of an anticholinergic syndrome after treatment with dimenhydrinate are described. The drug was
F J Forno et al.
The Journal of the American Osteopathic Association, 98(9), 502-504 (1998-10-24)
Ingestion of jimsonweed can result in hallucinations. Despite this side effect, the differential diagnosis of jimsonweed toxicity can be difficult, in part, because routine drug screens do not detect this agent. Such a delay in diagnosis not only postpones treatment
H Jesionowska et al.
The Journal of reproductive medicine, 36(3), 167-169 (1991-03-01)
A subcutaneous injection of physostigmine was employed to treat paraplegic anejaculation. The sperm motility was greater than in previously reported studies in which electroejaculation techniques were used for seminal emission.
A Bringmann
Neuroscience letters, 202(1-2), 13-16 (1995-12-29)
In previous experiments it was found that physostigmine application in unrestrained rats delayed the peak latency of the visual evoked potential (VEP). The present study was carried out to find the putative site that cholinergically mediates the latency delay of
C Lindén et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 115(7), 857-861 (1997-07-01)
To investigate if a pronounced ciliary muscle contraction, induced by physostigmine salicylate, can abolish the ocular hypotensive effect of latanoprost, a prostaglandin analogue, via inhibition of the uveoscleral outflow. A randomized, crossover study that was double-masked for latanoprost. Physostigmine was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service