Skip to Content
Merck
All Photos(1)

Key Documents

454788

Sigma-Aldrich

4-Iodo-2-methylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(CH3)NH2
CAS Number:
Molecular Weight:
233.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

86-89 °C (lit.)

SMILES string

Cc1cc(I)ccc1N

InChI

1S/C7H8IN/c1-5-4-6(8)2-3-7(5)9/h2-4H,9H2,1H3

InChI key

BGKLFAQCHHCZRZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Iodo-2-methylaniline.
Luo W, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 64(3), 0591-0591 (2008)
Synthesis of laterally substituted bistolane liquid crystals.
Hsu C-S, et al.
Liq. Cryst., 27(2), 283-287 (2000)
Synthesis and Fluorescence properties of Schiff bases of 4-hydroxy- 3-methoxy-5-nitrobenzaldehyde Journal of Chemical and Pharmaceutical Research.
Thorat B R, et al.
Journal of Chemistry, 3(6), 1045-1054 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service