Skip to Content
Merck
All Photos(1)

Key Documents

453471

Sigma-Aldrich

4-Cyanophenylhydrazine hydrochloride

97%

Synonym(s):

4-Hydrazinobenzonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4NHNH2 · HCl
CAS Number:
Molecular Weight:
169.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

241-244 °C (dec.) (lit.)

SMILES string

Cl.NNc1ccc(cc1)C#N

InChI

1S/C7H7N3.ClH/c8-5-6-1-3-7(10-9)4-2-6;/h1-4,10H,9H2;1H

InChI key

UXDLLFIRCVPPQP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Kostecka
Molecules (Basel, Switzerland), 17(3), 3560-3573 (2012-03-24)
In view of the growing demand for new compounds showing biological activity against pathogenic microorganisms, such as pathogenic and phytopathogenic fungi, the objective of this study was to synthesize a new group of aliphatic and aromatic derivatives of hydrazide. In
Synthesis and Structure-Activity Relationship Studies of Derivatives of the Dual Aromatase-Sulfatase Inhibitor 4-{[(4-Cyanophenyl)(4H-1,2,4-triazol-4-yl) amino] methyl} phenyl sulfamate.
Woo LW, et al.
ChemMedChem, 8(5), 779-799 (2013)
Design and synthesis of new water-soluble tetrazolide derivatives of celecoxib and rofecoxib as selective cyclooxygenase-2 (COX-2) inhibitors.
Navidpour L, et al.
Bioorganic & Medicinal Chemistry Letters, 16(17), 4483-4487 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service